(S)-1-Boc-2-propylpiperazine hydrochloride

Lieferant: Apollo Scientific

Synonyme: (S)-tert-Butyl 2-propylpiperazine-1-carboxylate hydrochloride

OR470454-1G
APOSOR470454-1GEA 112 EUR
APOSOR470454-1G
(S)-1-Boc-2-propylpiperazine hydrochloride
(S)-1-Boc-2-propylpiperazine hydrochloride
Formel: C₁₂H₂₄N₂O₂·ClH
Molecular Weight: 264,792 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11101260
CAS-Nummer: 1217478-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR