1-Allyl-7-methoxy-2-tetralone

Lieferant: Apollo Scientific

OR470455-5G
APOSOR470455-5GEA 1010 EUR
APOSOR470455-5G
1-Allyl-7-methoxy-2-tetralone
1-Allyl-7-methoxy-2-tetralone
Formel: C₁₄H₁₆O₂
Molecular Weight: 216,276 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21364428
CAS-Nummer: 29093-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR