Methyl 1-Cbz-3-allylpiperidine-3-carboxylate

Lieferant: Apollo Scientific

OR470458-5G
APOSOR470458-5GEA 489 EUR
APOSOR470458-5G
Methyl 1-Cbz-3-allylpiperidine-3-carboxylate
Methyl 1-Cbz-3-allylpiperidine-3-carboxylate
Formel: C₁₈H₂₃NO₄
Molecular Weight: 317,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641323
CAS-Nummer: 1363166-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR