1-Cbz-3-allylpiperidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR470459-5G
APOSOR470459-5GEA 1010 EUR
APOSOR470459-5G
1-Cbz-3-allylpiperidine-3-carboxylic acid
1-Cbz-3-allylpiperidine-3-carboxylic acid
Formel: C₁₇H₂₁NO₄
Molecular Weight: 303,353 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641324
CAS-Nummer: 1363166-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR