2-Ethyl-4-methylbenzoxazole

Lieferant: Apollo Scientific

OR470460-5G
APOSOR470460-5GEA 916 EUR
APOSOR470460-5G
2-Ethyl-4-methylbenzoxazole
2-Ethyl-4-methylbenzoxazole
Formel: C₁₀H₁₁NO
Molecular Weight: 161,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641330
CAS-Nummer: 1363166-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR