5-(tert-Butyl)benzoxazole

Lieferant: Apollo Scientific

OR470461-5G
APOSOR470461-5GEA 408 EUR
APOSOR470461-5G
5-(tert-Butyl)benzoxazole
5-(tert-Butyl)benzoxazole
Formel: C₁₁H₁₃NO
Molecular Weight: 175,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027424
CAS-Nummer: 908011-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR