5-(tert-Butyl)-2-ethylbenzoxazole

Lieferant: Apollo Scientific

OR470462-5G
APOSOR470462-5GEA 408 EUR
APOSOR470462-5G
5-(tert-Butyl)-2-ethylbenzoxazole
5-(tert-Butyl)-2-ethylbenzoxazole
Formel: C₁₃H₁₇NO
Molecular Weight: 203,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19375298
CAS-Nummer: 1267427-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR