2-Ethylbenzoxazole-6-carboxylic acid

Lieferant: Apollo Scientific

OR470463-5G
APOSOR470463-5GEA 916 EUR
APOSOR470463-5G
2-Ethylbenzoxazole-6-carboxylic acid
2-Ethylbenzoxazole-6-carboxylic acid
Formel: C₁₀H₉NO₃
Molecular Weight: 191,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11890765
CAS-Nummer: 501918-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR