2-Ethyl-5-methylbenzoxazole

Lieferant: Apollo Scientific

OR470466-5G
APOSOR470466-5GEA 611 EUR
APOSOR470466-5G
2-Ethyl-5-methylbenzoxazole
2-Ethyl-5-methylbenzoxazole
Formel: C₁₀H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD19374490
CAS-Nummer: 20514-29-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR