7-Bromo-2-ethylbenzoxazole

Lieferant: Apollo Scientific

OR470468-5G
APOSOR470468-5GEA 1120 EUR
APOSOR470468-5G
7-Bromo-2-ethylbenzoxazole
7-Bromo-2-ethylbenzoxazole
Formel: C₉H₈BrNO
Molecular Weight: 226.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19376426
CAS-Nummer: 1267428-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR