Diethyl 3-(tert-Butyldimethylsilyloxy)glutarate

Lieferant: Apollo Scientific

OR470469-25G OR470469-5G
APOSOR470469-25GEA 287 EUR
APOSOR470469-25G APOSOR470469-5G
Diethyl 3-(tert-Butyldimethylsilyloxy)glutarate
Diethyl 3-(tert-Butyldimethylsilyloxy)glutarate
Formel: C₁₅H₃₀O₅Si
Lagertemperatur: Raumtemperatur
MDL: MFCD16251279
CAS-Nummer: 91424-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR