4-(2-Chlorophenyl)-1,2,3-thiadiazole-5-carbonitrile

Lieferant: Apollo Scientific

OR470471-1G
APOSOR470471-1GEA 448 EUR
APOSOR470471-1G
4-(2-Chlorophenyl)-1,2,3-thiadiazole-5-carbonitrile
4-(2-Chlorophenyl)-1,2,3-thiadiazole-5-carbonitrile
Formel: C₉H₄ClN₃S
Molecular Weight: 221.666 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641340
CAS-Nummer: 265326-54-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR