Methyl 2-Isopropylbenzoate
Lieferant: Apollo Scientific
OR470473-1G
APOSOR470473-1GEA
448
EUR
APOSOR470473-1G
Methyl 2-Isopropylbenzoate
Methyl 2-Isopropylbenzoate
Formel:
C₁₁H₁₄O₂ Siedepunkt: 249 °C (760 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD20484275 CAS-Nummer: 6623-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...