Methyl 2,5-Dichloro-3-nitrobenzoate

Lieferant: Apollo Scientific

OR470475-5G
APOSOR470475-5GEA 1010 EUR
APOSOR470475-5G
Methyl 2,5-Dichloro-3-nitrobenzoate
Methyl 2,5-Dichloro-3-nitrobenzoate
Formel: C₈H₅Cl₂NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01529637
CAS-Nummer: 34408-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR