4-(Chloromethyl)phenylacetic acid

Lieferant: Apollo Scientific

OR470476-1G
APOSOR470476-1GEA 507 EUR
APOSOR470476-1G
4-(Chloromethyl)phenylacetic acid
4-(Chloromethyl)phenylacetic acid
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038831
CAS-Nummer: 56066-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR