4,5-Dihydro-1H-pyrido[2,3-b][1,4]diazepin-2(3H)-one

Lieferant: Apollo Scientific

OR470479-1G
APOSOR470479-1GEA 601 EUR
APOSOR470479-1G
4,5-Dihydro-1H-pyrido[2,3-b][1,4]diazepin-2(3H)-one
4,5-Dihydro-1H-pyrido[2,3-b][1,4]diazepin-2(3H)-one
Formel: C₈H₉N₃O
Molecular Weight: 163,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14705116
CAS-Nummer: 757966-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR