(1R,6S)-3-Methyl-3,8-diazabicyclo[4.2.0]octane

Lieferant: Apollo Scientific

OR470483-250MG
APOSOR470483-250MGEA 2020 EUR
APOSOR470483-250MG
(1R,6S)-3-Methyl-3,8-diazabicyclo[4.2.0]octane
(1R,6S)-3-Methyl-3,8-diazabicyclo[4.2.0]octane
Formel: C₇H₁₄N₂
Molecular Weight: 126,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205854
CAS-Nummer: 1434126-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR