cis-3-Methyl-3,8-diazabicyclo[4.2.0]octane
Lieferant: Apollo Scientific
OR470488-250MG
APOSOR470488-250MGEA
0
EUR
APOSOR470488-250MG
cis-3-Methyl-3,8-diazabicyclo[4.2.0]octane
cis-3-Methyl-3,8-diazabicyclo[4.2.0]octane
Formel:
C₇H₁₄N₂ Molecular Weight: 126,199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22205861 CAS-Nummer: 2135332-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...