cis-3-Methyl-3,7-diazabicyclo[4.2.0]octane

Lieferant: Apollo Scientific

OR470490-250MG
APOSOR470490-250MGEA 1940 EUR
APOSOR470490-250MG
cis-3-Methyl-3,7-diazabicyclo[4.2.0]octane
cis-3-Methyl-3,7-diazabicyclo[4.2.0]octane
Formel: C₇H₁₄N₂
Molecular Weight: 126,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205863
CAS-Nummer: 1932519-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR