Ethyl 2-Bromopyrrole-1-carboxylate

Lieferant: Apollo Scientific

OR470491-5G
APOSOR470491-5GEA 651 EUR
APOSOR470491-5G
Ethyl 2-Bromopyrrole-1-carboxylate
Ethyl 2-Bromopyrrole-1-carboxylate
Formel: C₇H₈BrNO₂
Molecular Weight: 218.048 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205865
CAS-Nummer: 1375064-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR