Ethyl 3-Bromoindole-1-carboxylate
Lieferant: Apollo Scientific
OR470492-5G
APOSOR470492-5GEA
510
EUR
APOSOR470492-5G
Ethyl 3-Bromoindole-1-carboxylate
Ethyl 3-Bromoindole-1-carboxylate
Formel:
C₁₁H₁₀BrNO₂ Molecular Weight: 268,107 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22205866 CAS-Nummer: 1375064-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...