1-Acetyl-3-bromo-5-chloroindole

Lieferant: Apollo Scientific

OR470493-5G
APOSOR470493-5GEA 713 EUR
APOSOR470493-5G
1-Acetyl-3-bromo-5-chloroindole
1-Acetyl-3-bromo-5-chloroindole
Formel: C₁₀H₇BrClNO
Molecular Weight: 272,526 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205870
CAS-Nummer: 1375064-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR