3-Bromo-5-chloro-1-(p-toluenesulphonyl)indole

Lieferant: Apollo Scientific

OR470495-5G
APOSOR470495-5GEA 713 EUR
APOSOR470495-5G
3-Bromo-5-chloro-1-(p-toluenesulphonyl)indole
3-Bromo-5-chloro-1-(p-toluenesulphonyl)indole
Formel: C₁₅H₁₁BrClNO₂S
Molecular Weight: 384,675 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205872
CAS-Nummer: 887338-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR