3-Bromo-4-chloro-1-(p-toluenesulphonyl)indole

Lieferant: Apollo Scientific

OR470497-5G
APOSOR470497-5GEA 713 EUR
APOSOR470497-5G
3-Bromo-4-chloro-1-(p-toluenesulphonyl)indole
3-Bromo-4-chloro-1-(p-toluenesulphonyl)indole
Formel: C₁₅H₁₁BrClNO₂S
Molecular Weight: 384,675 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205875
CAS-Nummer: 887338-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR