1-Acetyl-3-bromo-7-methylindole

Lieferant: Apollo Scientific

OR470498-5G
APOSOR470498-5GEA 510 EUR
APOSOR470498-5G
1-Acetyl-3-bromo-7-methylindole
1-Acetyl-3-bromo-7-methylindole
Formel: C₁₁H₁₀BrNO
Molecular Weight: 252,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205876
CAS-Nummer: 1375064-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR