3-Bromo-7-methyl-1-(p-toluenesulphonyl)indole

Lieferant: Apollo Scientific

OR470500-5G
APOSOR470500-5GEA 510 EUR
APOSOR470500-5G
3-Bromo-7-methyl-1-(p-toluenesulphonyl)indole
3-Bromo-7-methyl-1-(p-toluenesulphonyl)indole
Formel: C₁₆H₁₄BrNO₂S
Molecular Weight: 364,257 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205878
CAS-Nummer: 887338-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR