Methyl 4-morpholino-2-nitrobenzoate

Lieferant: Apollo Scientific

OR470502-5G
APOSOR470502-5GEA 561 EUR
APOSOR470502-5G
Methyl 4-morpholino-2-nitrobenzoate
Methyl 4-morpholino-2-nitrobenzoate
Formel: C₁₂H₁₄N₂O₅
Molecular Weight: 266,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205882
CAS-Nummer: 404010-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR