5-Propylpyrazole-3-carboxamide

Lieferant: Apollo Scientific

OR470503-5G
APOSOR470503-5GEA 561 EUR
APOSOR470503-5G
5-Propylpyrazole-3-carboxamide
5-Propylpyrazole-3-carboxamide
Formel: C₇H₁₁N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD16619745
CAS-Nummer: 957129-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR