4,4-Dimethylcyclohexanamine hydrochloride

Lieferant: Apollo Scientific

OR470504-5G
APOSOR470504-5GEA 1410 EUR
APOSOR470504-5G
4,4-Dimethylcyclohexanamine hydrochloride
4,4-Dimethylcyclohexanamine hydrochloride
Formel: C₈H₁₇N·ClH
Molecular Weight: 163,688 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08460047
CAS-Nummer: 25834-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR