α-(2-Methylsulphonylphenyl)benzylamine

Lieferant: Apollo Scientific

OR470505-1G
APOSOR470505-1GEA 713 EUR
APOSOR470505-1G
α-(2-Methylsulphonylphenyl)benzylamine
α-(2-Methylsulphonylphenyl)benzylamine
Formel: C₁₄H₁₅NO₂S
Molecular Weight: 261,339 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22574975
CAS-Nummer: 1408058-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR