α-(3-Biphenylyl)benzylamine

Lieferant: Apollo Scientific

OR470506-1G
APOSOR470506-1GEA 601 EUR
APOSOR470506-1G
α-(3-Biphenylyl)benzylamine
α-(3-Biphenylyl)benzylamine
Formel: C₁₉H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD22574976
CAS-Nummer: 1138152-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR