3-[(Diethylamino)methyl]phenylboronic acid

Lieferant: Apollo Scientific

OR470507-1G
APOSOR470507-1GEA 507 EUR
APOSOR470507-1G
3-[(Diethylamino)methyl]phenylboronic acid
3-[(Diethylamino)methyl]phenylboronic acid
Formel: C₁₁H₁₈BNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14687157
CAS-Nummer: 944483-39-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR