α-Acetoxy-(3,4-dimethoxyphenyl)acetonitrile

Lieferant: Apollo Scientific

OR470508-5G
APOSOR470508-5GEA 601 EUR
APOSOR470508-5G
α-Acetoxy-(3,4-dimethoxyphenyl)acetonitrile
α-Acetoxy-(3,4-dimethoxyphenyl)acetonitrile
Formel: C₁₂H₁₃NO₄
Molecular Weight: 235,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20128188
CAS-Nummer: 13782-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR