Methyl 5-(4-pyridinyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR470510-5G
APOSOR470510-5GEA 916 EUR
APOSOR470510-5G
Methyl 5-(4-pyridinyl)pyrazole-3-carboxylate
Methyl 5-(4-pyridinyl)pyrazole-3-carboxylate
Formel: C₁₀H₉N₃O₂
Molecular Weight: 203,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12188850
CAS-Nummer: 144252-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR