4-Chloro-7-(triisopropylsilyl)-7H-pyrrolo[2,3-d]pyrimidine
Lieferant: Apollo Scientific
OR470511-5G
APOSOR470511-5GEA
550
EUR
APOSOR470511-5G
4-Chloro-7-(triisopropylsilyl)-7H-pyrrolo[2,3-d]pyrimidine
4-Chloro-7-(triisopropylsilyl)-7H-pyrrolo[2,3-d]pyrimidine
Formel:
C₁₅H₂₄ClN₃Si Molecular Weight: 309,91 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22375698 CAS-Nummer: 870706-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...