Ethyl 5-(4-Pyridinyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR470512-5G
APOSOR470512-5GEA 916 EUR
APOSOR470512-5G
Ethyl 5-(4-Pyridinyl)pyrazole-3-carboxylate
Ethyl 5-(4-Pyridinyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₁N₃O₂
Molecular Weight: 217,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07783845
CAS-Nummer: 911461-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR