2-(Methylsulphonyl)acetamide

Lieferant: Apollo Scientific

OR470513-5G
APOSOR470513-5GEA 713 EUR
APOSOR470513-5G
2-(Methylsulphonyl)acetamide
2-(Methylsulphonyl)acetamide
Formel: C₃H₇NO₃S
Molecular Weight: 137,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025524
CAS-Nummer: 66913-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR