Ethyl 4-Benzyl-5-oxomorpholine-3-carboxylate

Lieferant: Apollo Scientific

OR470514-5G
APOSOR470514-5GEA 499 EUR
APOSOR470514-5G
Ethyl 4-Benzyl-5-oxomorpholine-3-carboxylate
Ethyl 4-Benzyl-5-oxomorpholine-3-carboxylate
Formel: C₁₄H₁₇NO₄
Molecular Weight: 263.289 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18252232
CAS-Nummer: 106910-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR