α-(4-Benzyloxyphenyl)benzylamine

Lieferant: Apollo Scientific

OR470515-5G
APOSOR470515-5GEA 804 EUR
APOSOR470515-5G
α-(4-Benzyloxyphenyl)benzylamine
α-(4-Benzyloxyphenyl)benzylamine
Formel: C₂₀H₁₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD22375699
CAS-Nummer: 748769-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR