Methyl 1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylate
Lieferant: Apollo Scientific
OR470516-5G
APOSOR470516-5GEA
1010
EUR
APOSOR470516-5G
Methyl 1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylate
Formel:
C₁₆H₂₀O₅ Molecular Weight: 292,327 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22423114 CAS-Nummer: 1385694-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...