Methyl 1-(3-Chlorophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470517-5G
APOSOR470517-5GEA 1010 EUR
APOSOR470517-5G
Methyl 1-(3-Chlorophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(3-Chlorophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₄H₁₅ClO₃
Molecular Weight: 266,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423116
CAS-Nummer: 1384264-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR