1-(2-Bromo-5-chlorophenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470518-5G
APOSOR470518-5GEA 1120 EUR
APOSOR470518-5G
1-(2-Bromo-5-chlorophenyl)-4-oxocyclohexanecarboxylic acid
1-(2-Bromo-5-chlorophenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₃H₁₂BrClO₃
Molecular Weight: 331,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423117
CAS-Nummer: 1385694-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR