1-(4-Nitrophenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470519-1G
APOSOR470519-1GEA 713 EUR
APOSOR470519-1G
1-(4-Nitrophenyl)-4-oxocyclohexanecarboxylic acid
1-(4-Nitrophenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₃H₁₃NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD22574977
CAS-Nummer: 1408074-40-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR