1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470520-1G
APOSOR470520-1GEA 713 EUR
APOSOR470520-1G
1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylic acid
1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₃H₁₂Cl₂O₃
Molecular Weight: 287,139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22574978
CAS-Nummer: 1408058-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR