1-(4-tert-Butylphenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470521-5G
APOSOR470521-5GEA 1010 EUR
APOSOR470521-5G
1-(4-tert-Butylphenyl)-4-oxocyclohexanecarboxylic acid
1-(4-tert-Butylphenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₇H₂₂O₃
Molecular Weight: 274,355 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423119
CAS-Nummer: 1385694-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR