Methyl 1-(2-Bromo-5-chlorophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470522-5G
APOSOR470522-5GEA 1120 EUR
APOSOR470522-5G
Methyl 1-(2-Bromo-5-chlorophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2-Bromo-5-chlorophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₄H₁₄BrClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22423120
CAS-Nummer: 1385694-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR