Methyl 1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470523-5G
APOSOR470523-5GEA 1010 EUR
APOSOR470523-5G
Methyl 1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₄H₁₄Cl₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22574979
CAS-Nummer: 1408058-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR