Methyl 1-(4-tert-Butylphenyl)-4-oxocyclohexanecarboxylate
Lieferant: Apollo Scientific
OR470524-5G
APOSOR470524-5GEA
1010
EUR
APOSOR470524-5G
Methyl 1-(4-tert-Butylphenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(4-tert-Butylphenyl)-4-oxocyclohexanecarboxylate
Formel:
C₁₈H₂₄O₃ Molecular Weight: 288,381 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22423122 CAS-Nummer: 1385694-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...