1-(3-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470525-5G
APOSOR470525-5GEA 1010 EUR
APOSOR470525-5G
1-(3-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
1-(3-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₄H₁₆O₄
Molecular Weight: 248,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20257290
CAS-Nummer: 51414-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR