1-(2-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470527-5G
APOSOR470527-5GEA 1010 EUR
APOSOR470527-5G
1-(2-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
1-(2-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₄H₁₆O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22423128
CAS-Nummer: 1385694-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR